Learn More
Trajan Scientific DIAMOND HEADSPACE SYRINGE
SDP
Supplier:  Trajan Scientific 008655
Encompass_Preferred
Trajan Diamond Headspace Syringes for CTC PAL RTC (Robotic Tool Change), Volume: 2500 uL, Model: 2.5MF-RTC/RSH-EGT-HS-6.5/0.63H, Length Needle: 65 mm, 2.5 in., Needle Gauge: 23 G, Connection Type: Fixed, Diameter Outer Needle: 0.024 in., 0.63 mm, For Use With: Gas chromatography, CTC PAL RTC
Catalog No. 50-253-0300