Learn More
Trajan Scientific DIAMOND HEADSPACE SYRINGE
SDP
Supplier:  Trajan Scientific 008631
Encompass_Preferred
SGE PAL Gas Tight Headspace Autosampler Syringes with 56mm Fixed Needle, Volume: 2.5 mL, Diameter Outer Needle: 0.63 mm, Needle Gauge: 23 G, Length Needle: 56 mm, Connection Type: Fixed, For Use With: Gas chromatography, CTC Autosampler, CombiPAL, PAL, LEAP, Model: 2.5MF-CTC-EGT-HS-5.6/0.63H
Catalog No. 50-253-0301