Learn More
Electron Microscopy Sciences DiATOME AFM Diamond Knife Cryo, 2.0 mm, 35° Knife angle
SDP
Catalog No. 501898737
Encompass
The high quality Diatome diamond knives are perfect for surfacing sectioning of biological and industrial specimens for investigation using AFM. In order to achieve the best results, only the highest quality diamond knives should be used. Diatome ultra AFM and cryo AFM knives are specially tested to ensure that they meet the increased quality requirements of AFM investigation. They produce extremely smooth sample surfaces and guarantee the best possible structure preservation.
Size: 2.0 mm
Knife angle: 35°
Catalog No. 50-189-8737 Supplier Electron Microscopy Sciences Supplier No. 20AFMCDL
The high quality Diatome diamond knives are perfect for surfacing sectioning of biological and industrial specimens for investigation using AFM. In order to achieve the best results, only the highest quality diamond knives should be used. Diatome ultra AFM and cryo AFM knives are specially tested to ensure that they meet the increased quality requirements of AFM investigation. They produce extremely smooth sample surfaces and guarantee the best possible structure preservation.
Size: 2.0 mm
Knife angle: 35°