Learn More
Electron Microscopy Sciences EMS Diamond Knife Cleaning Kit
SDP
Catalog No. 50930408
Encompass_Preferred
Consists of 5x5 ml vials of cleaning solution, 4 rods for cleaning the knife edge, and 1 hair curl for the removal of sections from the knife edge and the trough.
Catalog No. 50-930-408 Supplier Electron Microscopy Sciences Supplier No. 70600
Consists of 5x5 ml vials of cleaning solution, 4 rods for cleaning the knife edge, and 1 hair curl for the removal of sections from the knife edge and the trough.