Learn More
Electron Microscopy Sciences DiATOME Cryo Diamond Knife Dry, 1.8 mm, 35° Knife angle
SDP
Catalog No. 501898726
Encompass
Diatome cryo diamond knives are used for all low temperature situations such as sectioning of cryo-protected specimens, frozen hydrated specimens, and industrial samples such as polymers and rubber. Compared to glass knives cryo diamond knives produce ultrathin and semithin sections with the same knife, produce thinner sections, easily produce long ribbons of sections, and have less compression and greater uniformity of sections.
Size: 1.8 mm
Knife angle: 35°
Catalog No. 50-189-8726 Supplier Electron Microscopy Sciences Supplier No. 18CDL
Diatome cryo diamond knives are used for all low temperature situations such as sectioning of cryo-protected specimens, frozen hydrated specimens, and industrial samples such as polymers and rubber. Compared to glass knives cryo diamond knives produce ultrathin and semithin sections with the same knife, produce thinner sections, easily produce long ribbons of sections, and have less compression and greater uniformity of sections.
Size: 1.8 mm
Knife angle: 35°