Learn More
Electron Microscopy Sciences DiATOME Histo Diamond Knife 8.0 mm, 45° Knife angle
SDP
Catalog No. 501900063
Encompass
Diamond knives are for sectioning of hard and soft biological and industrial materials, non embedded or embedded in methacrylate or epoxy resins at room temperature. They have a 45 degree angle. The knives may be used on all ultramicrotomes and microtomes with a retraction of the specimen in the return phase.
Advantages of histo diamond knives over glass knives are perfect sections, free of scores or compression, serial sections without knife change, thinner sections, and extreme durability of the cutting edge. As well as no knife making procedure. Diatome knives have a lifetime guarantee.
Size: 8.0 mm
Knife angle: 45°
Catalog No. 50-190-0063 Supplier Electron Microscopy Sciences Supplier No. 80HIS
Diamond knives are for sectioning of hard and soft biological and industrial materials, non embedded or embedded in methacrylate or epoxy resins at room temperature. They have a 45 degree angle. The knives may be used on all ultramicrotomes and microtomes with a retraction of the specimen in the return phase.
Advantages of histo diamond knives over glass knives are perfect sections, free of scores or compression, serial sections without knife change, thinner sections, and extreme durability of the cutting edge. As well as no knife making procedure. Diatome knives have a lifetime guarantee.
Size: 8.0 mm
Knife angle: 45°