Learn More
Electron Microscopy Sciences DiATOME Ultra ATS Diamond Knife
SDP
Catalog No. 501929114
Encompass
The Ultra ATS Diamond knife is perfect for placing sections on Si wafers to view under the SEM. The knife comes in 3.0mm size with 35 deg angle. Please see the Technical Data Sheet for an illustration showing the technique of placing sections on a wafer.
Catalog No. 50-192-9114 Supplier Electron Microscopy Sciences Supplier No. 30ULATS
Add to Cart
Add to Cart
Request a Quote
The Ultra ATS Diamond knife is perfect for placing sections on Si wafers to view under the SEM. The knife comes in 3.0mm size with 35 deg angle. Please see the Technical Data Sheet for an illustration showing the technique of placing sections on a wafer.