Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Jumbo 2.0 mm 35° Knife angle
SDP
Catalog No. 501898740
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.0 mm
Knife angle: 35°
Catalog No. 50-189-8740 Supplier Electron Microscopy Sciences Supplier No. 20ULJ
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.0 mm
Knife angle: 35°