Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Jumbo 2.0 mm 45° Knife angle
SDP
Catalog No. 501898742
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.0 mm
Knife angle: 45°
Catalog No. 50-189-8742 Supplier Electron Microscopy Sciences Supplier No. 20USJ
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 2.0 mm
Knife angle: 45°