Learn More
Electron Microscopy Sciences DiATOME Ultra Diamond Knife Jumbo 3.0 mm 45° Knife angle
SDP
Catalog No. 501898822
Encompass
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.0 mm
Knife angle: 45°
Catalog No. 50-189-8822 Supplier Electron Microscopy Sciences Supplier No. 30USJ
Add to Cart
Add to Cart
Request a Quote
The optimal thickness range for cutting with these knives is between 30-150 nm.
Size: 3.0 mm
Knife angle: 45°