Learn More
Electron Microscopy Sciences Diamond Lapping Film 12", Plain Backing, 15 µm Particles
SDP
Catalog No. 50997268
Encompass_Preferred
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: Plain
Particles: 15 µm
Catalog No. 50-997-268 Supplier Electron Microscopy Sciences Supplier No. 5035430
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: Plain
Particles: 15 µm