Learn More
Electron Microscopy Sciences Diamond Lapping Film 12", PSA Backing, 15 µm Particles
SDP
Catalog No. 50997277
Encompass_Preferred
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: PSA
Particles: 15 µm
Catalog No. 50-997-277 Supplier Electron Microscopy Sciences Supplier No. 5035530
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: PSA
Particles: 15 µm