Learn More
Electron Microscopy Sciences Diamond Lapping Film 12", PSA Backing, 30 µm Particles
SDP
Catalog No. 50997278
Encompass_Preferred
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: PSA
Particles: 30 µm
Catalog No. 50-997-278 Supplier Electron Microscopy Sciences Supplier No. 5035535
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: PSA
Particles: 30 µm