Learn More
Electron Microscopy Sciences Diamond Lapping Film 12", PSA Backing, 0.5 µm Particles
SDP
Catalog No. 50997272
Encompass_Preferred
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: PSA
Particles: 0.5 µm
Catalog No. 50-997-272 Supplier Electron Microscopy Sciences Supplier No. 5035505
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis.
Backing: PSA
Particles: 0.5 µm