Learn More
Trajan Scientific DIAMOND MS NEEDLES
SDP
Catalog No. 502530267
Encompass_Preferred
Trajan Diamond Thermo RSH GC Autosampler 10 to 50uL Syringes, Diameter Inner Needle: 0.24 mm, 0.009 in., Description: Needle coating: Ultra Smooth Hydrophobic, Length Needle: 57 mm, 2.24 in., Needle Gauge: 23 G, Volume: 10 to 50 uL, Diameter Outer Needle: 0.024 in., 0.63 mm
Catalog No. 50-253-0267 Supplier Trajan Scientific Supplier No. 0355620
Trajan Diamond Thermo RSH GC Autosampler 10 to 50uL Syringes, Diameter Inner Needle: 0.24 mm, 0.009 in., Description: Needle coating: Ultra Smooth Hydrophobic, Length Needle: 57 mm, 2.24 in., Needle Gauge: 23 G, Volume: 10 to 50 uL, Diameter Outer Needle: 0.024 in., 0.63 mm