Learn More
Trajan Scientific DIAMOND MS SYRINGE
SDP
Supplier:  Trajan Scientific 002790
Encompass_Preferred
Trajan Diamond MS Thermo RSH and CTC RTC Autosampler Syringes, Volume: 10 uL, Model: 10R-RTC/RSH-GT-MS1, Connection Type: Removable, For Use With: Gas/Liquid chromatography, Thermo RSH and CTC RTC, Max. Temperature: 662 deg.F, 350 deg.C, Quantity: Each
Catalog No. 50-253-0392