Learn More
Trajan Scientific DIAMOND MS SYRINGE
SDP
Supplier:  Trajan Scientific 005750
Encompass_Preferred
Trajan Diamond MS Manual Syringes, Volume: 100 uL, Model: 100R-GT-MS1, Connection Type: Removable, Includes: Removable ultra smooth needle, Quantity: Each
Catalog No. 50-253-0326