Learn More
Trajan Scientific DIAMOND MS SYRINGE
SDP
Catalog No. 502530393
Encompass_Preferred
Trajan Diamond MS CTC PAL Autosampler Syringes, Volume: 10 uL, Model: 10R-CTC-GT-MS1, Connection Type: Removable, For Use With: Gas/Liquid chromatography, CTC PAL autosampler syringes, Max. Temperature: 662 deg.F, 350 deg.C, Needle Point Style: Plunger, Quantity: Each, Diamond MS Syringe
Catalog No. 50-253-0393 Supplier Trajan Scientific Supplier No. 002770
Trajan Diamond MS CTC PAL Autosampler Syringes, Volume: 10 uL, Model: 10R-CTC-GT-MS1, Connection Type: Removable, For Use With: Gas/Liquid chromatography, CTC PAL autosampler syringes, Max. Temperature: 662 deg.F, 350 deg.C, Needle Point Style: Plunger, Quantity: Each, Diamond MS Syringe