Learn More
Electron Microscopy Sciences Glennel Diamond Paste UB Water/Oil Soluble, Monocrystalline 0.1 µm 5 GR
SDP
Catalog No. 50997314
Encompass_Preferred
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. Excellent for most applications.
Type: Monocrystalline
Size: 0.1 µm
Catalog No. 50-997-314 Supplier Electron Microscopy Sciences Supplier No. 5037010
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. Excellent for most applications.
Type: Monocrystalline
Size: 0.1 µm