Learn More
Electron Microscopy Sciences Glennel Diamond Paste UBS Water/Oil Soluble, Polycrystalline 0.1 µm 5 GR
SDP
Catalog No. 50997315
Encompass_Preferred
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. Excellent for most applications. used for microscopy.
Type: Polycrystalline
Size: 0.1 µm
Catalog No. 50-997-315 Supplier Electron Microscopy Sciences Supplier No. 5037011
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. Excellent for most applications. used for microscopy.
Type: Polycrystalline
Size: 0.1 µm