Learn More
Electron Microscopy Sciences Glennel Diamond Paste UBS Water/Oil Soluble, Polycrystalline 0.5 µm 5 GR
SDP
Catalog No. 50997319
Encompass_Preferred
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. UB formulation is excellent for most applications and used for polishing laboratory specimens. Polycrystalline diamonds are ideal for microscopy, materialography, and where surface finish is extremely critical.
Type: Polycrystalline
Size: 0.5 µm
Catalog No. 50-997-319 Supplier Electron Microscopy Sciences Supplier No. 5037031
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. UB formulation is excellent for most applications and used for polishing laboratory specimens. Polycrystalline diamonds are ideal for microscopy, materialography, and where surface finish is extremely critical.
Type: Polycrystalline
Size: 0.5 µm