Learn More
Electron Microscopy Sciences Glennel Diamond Paste UC Oil Soluble, Monocrystalline 0.5 µm 5 GR
SDP
Catalog No. 50997333
Encompass_Preferred
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. UC formulation is for fast cutting action and superior finish on wire dies, molds, cold heading dies, and more. Use with Formula C extender. Monocrystalline diamonds provide a cost effective means for good stock removal and finish.
Type: Monocrystalline
Size: 0.5 µm
Catalog No. 50-997-333 Supplier Electron Microscopy Sciences Supplier No. 5037130
Glennel diamond products are specially formulated to help you prepare surfaces that are usually difficult to polish and require precision finish. To satisfy today's demand the diamond compounds must be complex with near perfect tolerances as is ours. UC formulation is for fast cutting action and superior finish on wire dies, molds, cold heading dies, and more. Use with Formula C extender. Monocrystalline diamonds provide a cost effective means for good stock removal and finish.
Type: Monocrystalline
Size: 0.5 µm