Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampco Diamond Point Chisel
SDP

Catalog No. 19173562 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
8 in.
12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
19-173-562 8 in.
19-173-563 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-173-562 Supplier Ampco Supplier No. C6
Add to Cart
Add to Cart

Non-sparking, non-magnetic, and corrosion-resistant diamond point chisel. Made in the USA.

  • Sharp diamond-shaped tip easily reaches into narrow slots, corners, and tight spaces.
  • Offers detailed control compared to conventional flat chisels.
  • Safe for use in hazardous environments/workspaces.

Specifications

Product Type Diamond Point Chisel
Length (English) 8 in.
Diameter (Metric) 20 mm
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.