Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Inverted Wide Cone, 8.3 mm Tip, 083 I.S.O.
SDP
Catalog No. 5025444
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 8.3 mm
I.S.O. size: 083
Catalog No. 50-254-44 Supplier Electron Microscopy Sciences Supplier No. 6215583
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 8.3 mm
I.S.O. size: 083