Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Cone, 2.4 mm Tip, 024 I.S.O.
SDP
Catalog No. 5025437
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 2.4 mm
I.S.O. size: 024
Catalog No. 50-254-37 Supplier Electron Microscopy Sciences Supplier No. 6215524
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 2.4 mm
I.S.O. size: 024