Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Flat & Cylinder, 3.5 mm Tip, 035 I.S.O.
SDP
Catalog No. 5025443
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 3.5 mm
I.S.O. size: 035
Catalog No. 50-254-43 Supplier Electron Microscopy Sciences Supplier No. 6215535
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 3.5 mm
I.S.O. size: 035