Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Inverted Cone 2.3 mm Tip, 023 I.S.O.
SDP
Catalog No. 5025438
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 2.3 mm
I.S.O. size: 023
Catalog No. 50-254-38 Supplier Electron Microscopy Sciences Supplier No. 6215523
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 2.3 mm
I.S.O. size: 023