Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Large Round, 2.9 mm Tip, 029 I.S.O.
SDP
Catalog No. 5025442
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 2.9 mm
I.S.O. size: 029
Catalog No. 50-254-42 Supplier Electron Microscopy Sciences Supplier No. 6215529
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 2.9 mm
I.S.O. size: 029