Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Narrow Cone, 1.8 mm Tip, 018 I.S.O.
SDP
Catalog No. 5025439
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 1.8 mm
I.S.O. size: 018
Catalog No. 50-254-39 Supplier Electron Microscopy Sciences Supplier No. 6215518
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 1.8 mm
I.S.O. size: 018