Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Square Edge, 6.3 mm Tip, 063 I.S.O.
SDP
Catalog No. 5025441
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 6.3 mm
I.S.O. size: 063
Catalog No. 50-254-41 Supplier Electron Microscopy Sciences Supplier No. 6215563
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 6.3 mm
I.S.O. size: 063