Learn More
Electron Microscopy Sciences Flexible Shaft Machine Diamond Points Tapered Cone, 5.2 mm Tip, 052 I.S.O.
SDP
Catalog No. 5025445
Encompass_Preferred
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 5.2 mm
I.S.O. size: 052
Catalog No. 50-254-45 Supplier Electron Microscopy Sciences Supplier No. 6215552
All diamond points are mounted on 3/32" (2.35 mm) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Tip: 5.2 mm
I.S.O. size: 052