Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Diamond PosiStop LTS-Compatible Pipette Tips, Bulk

Catalog No. 23763010
Change view
Click to view available options
Volume (Metric):
20 μL
200 μL
1000 μL
Sterility:
Non-sterile
Sterile
Tip Style:
Low Retention
Standard
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Tip Style
23-763-010 1000 μL Non-sterile Standard
23-763-008 20 μL Non-sterile Standard
23-763-023 20 μL Sterile Standard
23-763-041 20 μL Sterile Low Retention
23-763-009 200 μL Non-sterile Standard
23-763-030 200 μL Sterile Standard
23-763-045 200 μL Sterile Low Retention
23-763-037 1000 μL Sterile Standard
23-763-049 1000 μL Sterile Low Retention
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 23-763-010 Supplier Globe Scientific Supplier No. 154035
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Globe Scientific PosiStop LTS-compatible Pipette Tips ensure a secure seal with a unique positive-stop collar and fail-safe design. Optimized for LTS pipettors, they enhance accuracy, deliver consistent results, and offer improved ergonomic comfort in high-throughput lab settings.

  • Designed for optimal performance with LTS pipettors, including Diamond PosiStop and Rainin LTS models.
  • Made from medical grade virgin polypropylene, free of any metal contaminants.
  • Rigorously tested and certified to be free from DNase, RNase, Pyrogens, PCR Inhibitors, and Endotoxins, ensuring the highest level of purity for critical experiments.
  • Manufactured in a state-of-the-art 100k cleanroom facility utilizing advanced automated processes.
  • Featuring color-coded inserts for quick and effortless identification and matching with Globe Diamond PosiStop pipettors.
  • All tips have graduation marks, providing a visual reference to reduce the chance of errors during sample aspiration.
  • Equipped with specially designed polyethylene (PE) filters that effectively prevent aerosols, significantly reducing the risk of cross-contamination.
  • Available in a variety of styles to meet specific requirements: Filter, Non-Filter, Standard, and Low Retention options.
  • Offered in multiple packaging formats: Rack, Bulk, Reload, and eco-friendly GreenRacks.

Specifications

Volume (Metric) 1000 μL
Filtered Non-filtered
Sterility Non-sterile
Color Neutral
Autoclavable Autoclavable
Format Bulk, Bagged
Tip Style Standard
Product Type Pipette Tip
Color Code White
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated Yes
Material Virgin Polypropylene
Fit for Purpose Certified Quality
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.