Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Globe Scientific Diamond PosiStop Pipettors, 12-Channel

Catalog No. 23763094
Change view
Click to view available options
Volume (Metric):
10 μL
20 μL
50 μL
100 μL
200 μL
300 μL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-763-094 10 μL
23-763-096 20 μL
23-763-099 50 μL
23-763-095 100 μL
23-763-097 200 μL
23-763-098 300 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 23-763-094 Supplier Globe Scientific Supplier No. 336510
Add to Cart
Add to Cart
This item is not returnable. View return policy

Diamond PosiStop 12-channel LTS pipettors deliver precision and comfort with a 360° rotating manifold and individually calibrated channels. ISO17025-certified and backed by a 3-year warranty, they're lightweight, ergonomic, and ideal for high-throughput, multichannel pipetting tasks.

  • Individually Calibrated Channels: Ensures consistent accuracy across all channels.
  • Removable Manifold: Fully autoclavable and UV resistant, offering flexibility and durability in sterilization procedures.
  • 3-Year Warranty: Affirms the quality and reliability of our pipettors.
  • Color-Coded Plungers: Facilitates easy volume identification, optimizing workflow efficiency.
  • Low Plunger Force: Provides a virtually fatigue-free pipetting experience.
  • Magnetically Assisted Piston: Delivers exceptionally high accuracy and consistency.
  • Ergonomic and Lightweight Design: Reduces strain and enhances comfort during prolonged use.
  • Precise Volume Adjustment: Allows for meticulous control, essential for accurate lab work.
  • Secure Locking Mechanism: Prevents unintentional volume alterations, ensuring dependable results.

Specifications

Compatible Tips Rainin LTS
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Red
Disposable Reusable
For Use With Pipette Tip 20 μL
Grip Type Ergonomic
Material PVDF, Stainless Steel
Number of Channels 12
Programmability No
Sterility Non-sterile
Type Adjustable Volume Pipette
Volume (Metric) 10 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.