Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamond powder, natural, 40-60 micron

Catalog No. AA3898403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3898403 1 g
AA3898406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3898403 Supplier Thermo Scientific Chemicals Supplier No. 03898403
Add to Cart

Diamond powder is generally used as an abrasive to grind and polish hard surfaces

Diamond powder is generally used as an abrasive to grind and polish hard surfaces.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diamond powder
Name Note Natural
Physical Form Powder
Quantity 1 g
Assay Percent Notes (metals basis)
MDL Number MFCD00211867
Percent Purity 99.9%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.