Alfa Aesar™ Diamond powder, synthetic, 40-60 micron, 99.9% (metals basis)

Diamond powder is generally used as an abrasive to grind and polish hard surfaces

Manufacturer: Alfa Aesar™ 1340203

Quantity 1g

 View more versions of this product

Catalog No. AA1340203

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD00211867
    Chemical Name or Material Diamond powder, Synthetic

    Specifications

    Quantity 1g
    Physical Form Powder
    MDL Number MFCD00211867
    Particle Size 40 to 60 Micron
    TSCA TSCA
    Chemical Name or Material Diamond powder, Synthetic
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit