Description
- One hand adjustment of tip volume
- Dosing volumes both of which are clearly indicated in vision pane on pipettor
- Accommodates 12 tip sizes and has 10 adjustments for 120 dispensing volume options
- Designed for Diamond RV-Pette PRO™ dispenser tips
- Compatible with syringe tips from most manufacturers, including Eppendorf™, Combitips™, Combitips Plus™, and Combitips Advanced™
- Maintenance free design requires no batteries or charging
- Materials are resistant to most chemicals
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 8655 |
| For Use With | Diamond RV-Pette PRO Dispenser Tip |
| Model | RV-Pette PRO |
| Number of Channels | 1 |
| Type | Repeating Pipette |
| Warranty | 1 Year |
| Weight (Metric) | 87 g |