Learn More
Structure Probe Inc DIAMOND SCRIBE
SDP
Catalog No. NC9531768
Encompass
SPI Diamond Scribe, High precision marking and scribing/scoring on silicon, glass and other hard surfaces. The standard product comes with a diamond 0.9mm (0.035 in. ) to 1.0mm (0.040 in. ) in diameter, with an exposed angle of 90-100 deg.
Catalog No. NC9531768 Supplier Structure Probe Inc Supplier No. 05022AB
SPI Diamond Scribe, High precision marking and scribing/scoring on silicon, glass and other hard surfaces. The standard product comes with a diamond 0.9mm (0.035 in. ) to 1.0mm (0.040 in. ) in diameter, with an exposed angle of 90-100 deg.