Learn More
Ted Pella Inc DIAMOND SCRIBE FINE NO 54467
SDP
Catalog No. NC9604633
Encompass
Fine Point Diamond Scriber Overall length is 120mm (4.72 in. ) diamond has a 60degree included angle Bent 30degree may also be used to clean or trim circuits or break or build up new lines
Catalog No. NC9604633 Supplier Ted Pella Inc Supplier No. 54467
Fine Point Diamond Scriber Overall length is 120mm (4.72 in. ) diamond has a 60degree included angle Bent 30degree may also be used to clean or trim circuits or break or build up new lines