Learn More
Ted Pella Inc DIAMOND SCRIBE STRAIGHT CUT
SDP
Catalog No. NC9983656
Encompass
Diamond Scriber, Head: Straight, Diamond Tip, Angle: 90 deg. Tip: Fine, Overall Length: 6-1/2 ft. (16.5cm), 1/EA
Catalog No. NC9983656 Supplier Ted Pella Inc Supplier No. 54484
Diamond Scriber, Head: Straight, Diamond Tip, Angle: 90 deg. Tip: Fine, Overall Length: 6-1/2 ft. (16.5cm), 1/EA