Learn More
Andwin Scientific DIAMOND SCRIBER
SDP
Catalog No. NC9432883
Encompass_Preferred
The Diamond Scriber can be utilized for making permanent markings on several kinds of surfaces. including glass slides, test tubes, and laboratory glassware. This product features a clear aluminum handle that provides comfort for users. The tips are retractable for safe utilization.
Catalog No. NC9432883 Supplier Andwin Scientific Supplier No. 52865005
The Diamond Scriber can be utilized for making permanent markings on several kinds of surfaces. including glass slides, test tubes, and laboratory glassware. This product features a clear aluminum handle that provides comfort for users. The tips are retractable for safe utilization.