Learn More
Electron Microscopy Sciences High Precision Diamond Scriber with Bent Heavy Duty Tip, 0.03" diameter, 0.12" Tip length
SDP
Catalog No. 50996731
Encompass
Precision scriber has a natural diamond tip mounted on 3 mm diameter tool cone.
Diameter: 0.03"
Tip length: 0.12"
Catalog No. 50-996-731 Supplier Electron Microscopy Sciences Supplier No. 62105HC
Precision scriber has a natural diamond tip mounted on 3 mm diameter tool cone.
Diameter: 0.03"
Tip length: 0.12"