Learn More
Electron Microscopy Sciences High Precision Diamond Scriber with Bent Tip, 0.02" diameter, 0.06" Tip length
SDP
Catalog No. 50996728
Encompass
Precision scriber has a natural diamond tip mounted on 3 mm diameter tool cone.
Diameter: 0.02"
Tip length: 0.06"
Catalog No. 50-996-728 Supplier Electron Microscopy Sciences Supplier No. 62105B1
Precision scriber has a natural diamond tip mounted on 3 mm diameter tool cone.
Diameter: 0.02"
Tip length: 0.06"