Learn More
Electron Microscopy Sciences High Precision Diamond Scribing Tools Complete Set, diameter, Tip length
SDP
Catalog No. 50996727
Encompass
A newly designed diamond scribing tool set containing eight precision natural diamond scribes in various sizes and configurations making it perfect for precision work in microscopy. The set consists of eight diamond tips each mounted on 3 mm in diameter interchangeable tool cone and an MT-1 tool handle. The tool and handle are all housed in a hardwood instrument case.
Catalog No. 50-996-727 Supplier Electron Microscopy Sciences Supplier No. 6210500
Add to Cart
Add to Cart
A newly designed diamond scribing tool set containing eight precision natural diamond scribes in various sizes and configurations making it perfect for precision work in microscopy. The set consists of eight diamond tips each mounted on 3 mm in diameter interchangeable tool cone and an MT-1 tool handle. The tool and handle are all housed in a hardwood instrument case.