Learn More
Electron Microscopy Sciences Diamond Scribing Tool Bent, 0.25 mm
SDP
Catalog No. 50996812
Encompass_Preferred
For use with Precision Pin Vise and Collets, 0.80mm (0.003") diameter mounting shank, 0.10" (0.25mm) diameter, 1.5mm tip length
Catalog No. 50-996-812 Supplier Electron Microscopy Sciences Supplier No. 6214002
For use with Precision Pin Vise and Collets, 0.80mm (0.003") diameter mounting shank, 0.10" (0.25mm) diameter, 1.5mm tip length