Learn More
Electron Microscopy Sciences Diamond Scribing Tool Bent, 0.50 mm
SDP
Catalog No. 50996816
Encompass_Preferred
Bent tip scriber, for use with Precision Pin Vise and Collets, 0.020" (0.50mm) diameter by 0.60" (1.5mm) tip length.
Catalog No. 50-996-816 Supplier Electron Microscopy Sciences Supplier No. 6214006
Bent tip scriber, for use with Precision Pin Vise and Collets, 0.020" (0.50mm) diameter by 0.60" (1.5mm) tip length.