Learn More
Electron Microscopy Sciences Diamond Scribing Tool Bent, 0.50 mm
SDP
Supplier:  Electron Microscopy Sciences 6214004
Encompass_Preferred
Bent extender tip scriber for use with Precision Pin Vise and Collets, 0.020" (0.50mm) diameter by 0.200"(5.0mm) tip length
Catalog No. 50-996-814