Learn More
Electron Microscopy Sciences PrecISIon Pin-Vise & Collet
SDP
Catalog No. 50996811
Encompass_Preferred
This high precision pin vise handle is machined from the finest material, nonrolling gold-anodized aluminum, which makes it lightweight and versatile. It comes complete with a precision drawbar for positive collets chuck clamping. It features an 8 mm (0.31") hexagonal barrel dimension by 100 mm (4") long (overall length is 115 mm fully assembled) and it has a knurled finger-grip for positive precision control for handling and tool manipulation.
The vice is supplied with a standard collet chuck opening of 2 mm (0.78") diameter and will hold a selection of our specially designed tools - and collets adapter 0.8 mm (0.003”) for diamond stylus chucking.
Catalog No. 50-996-811 Supplier Electron Microscopy Sciences Supplier No. 6214000
This high precision pin vise handle is machined from the finest material, nonrolling gold-anodized aluminum, which makes it lightweight and versatile. It comes complete with a precision drawbar for positive collets chuck clamping. It features an 8 mm (0.31") hexagonal barrel dimension by 100 mm (4") long (overall length is 115 mm fully assembled) and it has a knurled finger-grip for positive precision control for handling and tool manipulation.
The vice is supplied with a standard collet chuck opening of 2 mm (0.78") diameter and will hold a selection of our specially designed tools - and collets adapter 0.8 mm (0.003”) for diamond stylus chucking.