Description
- Simple plug and play operation
- Ergonomic, lightweight design helps reduce fatigue
- Powerful motors for fast liquid draw
- Precise control of coarse and fine speeds for speed and accuracy
- Accommodates all sizes of serological pipettes, glass and plastic
- 0.45 μm hydrophobic filter prevents contamination
- Long-life lithium ion battery for hours of use and no 'memory'
- Use while charging without any wait time
- Autoclavable nose cone ensures contamination-free pipetting
Specifications
Specifications
| Battery Type | Lithium-ion |
| Compatible Tips | All Serological Pipettes |
| Autoclavable | Not Autoclavable |
| Disposable | Reusable |
| Motor Type | Digital |
| Number of Speeds | 6 Speeds |