Learn More
Globe Scientific™ Diamond SeroFlow Pipette Controllers
Catalog No. p-7226629
Change view
Click to view available options
Motor Type:
Analog
Digital
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Motor Type
23-556-042 Analog
23-556-043 Digital
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-556-042 Supplier Globe Scientific™ Supplier No. 3385
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
This pipette controller has a comfortable, ergonomic design for hours of continuous use in the laboratory.
  • Simple plug and play operation
  • Ergonomic, lightweight design helps reduce fatigue
  • Powerful motors for fast liquid draw
  • Precise control of coarse and fine speeds for speed and accuracy
  • Accommodates all sizes of serological pipettes, glass and plastic
  • 0.45 μm hydrophobic filter prevents contamination
  • Long-life lithium ion battery for hours of use and no 'memory'
  • Use while charging without any wait time
  • Autoclavable nose cone ensures contamination-free pipetting
Specifications
Battery Type Lithium-ion
Compatible Tips All Serological Pipettes
Autoclavable Not Autoclavable
Disposable Reusable
Sterility Non-sterile
Motor Type Analog