Learn More
Sigma Aldrich Fine Chemicals Biosciences Diamond Shaped Weigh Boats | 30ml | 500ea
Catalog No. NC1750570
Encompass
These flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss. Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
Catalog No. NC1750570 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS1425B
These flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss. Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.